Assay Details
Gene Symbol: Cenps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004863607.2 | XP_004863664.1 | 2 - 3 | 361 | 66 | |
XM_004863610.2 | XP_004863667.1 | 4 - 5 | 412 | 66 | ||
XM_004863611.2 | XP_004863668.1 | 4 - 5 | 416 | 66 | ||
XM_004863612.2 | XP_004863669.1 | 3 - 4 | 321 | 66 | ||
XM_004863614.3 | XP_004863671.1 | 2 - 3 | 323 | 66 | ||
XM_021243626.1 | XP_021099285.1 | 3 - 4 | 339 | 66 | ||
XM_021243627.1 | XP_021099286.1 | 3 - 4 | 343 | 66 | ||
XM_021243628.1 | XP_021099287.1 | 2 - 3 | 249 | 66 | ||
XM_021243629.1 | XP_021099288.1 | 4 - 5 | 397 | 66 | ||
XM_021243630.1 | XP_021099289.1 | 3 - 4 | 306 | 66 | ||
XM_021243631.1 | XP_021099290.1 | 2 - 3 | 267 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004863610.2, XM_004863611.2, XM_004863612.2, XM_021243629.1, XM_021243630.1, XM_021243631.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
