Assay Details
Gene Symbol: Cenps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004863607.2 | XP_004863664.1 | 3 - 4 | 399 | 88 | |
XM_004863610.2 | XP_004863667.1 | 5 - 6 | 450 | 88 | ||
XM_004863611.2 | XP_004863668.1 | 5 - 6 | 454 | 88 | ||
XM_004863612.2 | XP_004863669.1 | 4 - 5 | 359 | 88 | ||
XM_004863614.3 | XP_004863671.1 | 3 - 4 | 361 | 88 | ||
XM_021243626.1 | XP_021099285.1 | 4 - 5 | 377 | 88 | ||
XM_021243627.1 | XP_021099286.1 | 4 - 5 | 381 | 88 | ||
XM_021243628.1 | XP_021099287.1 | 3 - 4 | 287 | 88 | ||
XM_021243629.1 | XP_021099288.1 | 5 - 6 | 435 | 88 | ||
XM_021243630.1 | XP_021099289.1 | 4 - 5 | 344 | 88 | ||
XM_021243631.1 | XP_021099290.1 | 3 - 4 | 305 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
