Assay Details
| Gene Symbol: Alg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845363.2 | XP_004845420.1 | 2 - 3 | 395 | 70 | |
| XM_013072591.2 | XP_012928045.1 | 2 - 3 | 395 | 70 | ||
| XM_021258769.1 | XP_021114428.1 | 2 - 3 | 395 | 70 | ||
More Information
Set Membership: |
|

Alg12
Genomic Map