Assay Details
Gene Symbol: Qser1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013074806.2 | XP_012930260.2 | 3 - 4 | 507 | 71 | |
XM_013074807.2 | XP_012930261.2 | 3 - 4 | 507 | 71 | ||
XM_021264389.1 | XP_021120048.1 | 2 - 3 | 183 | 71 | ||
XM_021264390.1 | XP_021120049.1 | 4 - 5 | 534 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
