Assay Details
| Gene Symbol: LOC101712296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835763.3 | XP_004835820.1 | 6 - 7 | 986 | 64 | |
| XM_004835764.3 | XP_004835821.1 | 6 - 7 | 913 | 64 | ||
| XM_004835765.3 | XP_004835822.1 | 6 - 7 | 912 | 64 | ||
| XM_021264141.1 | XP_021119800.1 | 6 - 7 | 912 | 64 | ||
More Information
Set Membership: |
|

LOC101712296
Genomic Map