Assay Details
| Gene Symbol: Ccdc92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843965.3 | XP_004844022.1 | 4 - 5 | 474 | 88 | |
| XM_004843966.2 | XP_004844023.1 | 3 - 4 | 263 | 88 | ||
| XM_021257630.1 | XP_021113289.1 | 4 - 5 | 467 | 88 | ||
| XM_021257631.1 | XP_021113290.1 | 4 - 5 | 327 | 88 | ||
More Information
Set Membership: |
|

Ccdc92
Genomic Map