Assay Details
Gene Symbol: Pdcd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013077458.2 | XP_012932912.1 | 1 - 2 | 150 | 63 | |
XM_021239754.1 | XP_021095413.1 | 1 - 2 | 151 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
