Assay Details
Gene Symbol: Pum3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838223.3 | XP_004838280.1 | 9 - 10 | 1307 | 72 | |
XM_004838226.3 | XP_004838283.1 | 8 - 9 | 928 | 72 | ||
XM_013069688.2 | XP_012925142.1 | 10 - 11 | 1489 | 72 | ||
XM_013069689.2 | XP_012925143.1 | 9 - 10 | 1236 | 72 | ||
XM_021252761.1 | XP_021108420.1 | 10 - 11 | 1363 | 72 | ||
XM_021252762.1 | XP_021108421.1 | 10 - 11 | 1292 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
