Assay Details
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004863166.2 | XP_004863223.1 | 10 - 11 | 1386 | 89 | |
XM_013079049.2 | XP_012934503.1 | 9 - 10 | 1282 | 89 | ||
XM_013079050.2 | XP_012934504.1 | 11 - 12 | 1447 | 89 | ||
XM_013079051.2 | XP_012934505.1 | 9 - 10 | 1261 | 89 | ||
XM_013079052.2 | XP_012934506.1 | 9 - 10 | 1287 | 89 | ||
XM_013079053.2 | XP_012934507.1 | 7 - 8 | 987 | 89 | ||
XM_021243145.1 | XP_021098804.1 | 9 - 10 | 1213 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
