Assay Details
Gene Symbol: Ints13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845732.2 | XP_004845789.1 | 6 - 7 | 1228 | 77 | |
XM_004845733.2 | XP_004845790.1 | 6 - 7 | 1228 | 77 | ||
XM_013072540.2 | XP_012927994.1 | 6 - 7 | 1228 | 77 | ||
XM_021259067.1 | XP_021114726.1 | 6 - 7 | 1229 | 77 | ||
XM_021259068.1 | XP_021114727.1 | 6 - 7 | 1228 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
