Assay Details
| Gene Symbol: Arl6ip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835938.2 | XP_004835995.1 | 2 - 3 | 459 | 104 | |
| XM_021266182.1 | XP_021121841.1 | 2 - 3 | 459 | 104 | ||
More Information
Set Membership: |
|

Arl6ip6
Genomic Map