Assay Details
Gene Symbol: Nsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847401.3 | XP_004847458.1 | 14 - 15 | 3567 | 109 | |
XM_013073021.2 | XP_012928475.1 | 15 - 16 | 3645 | 109 | ||
XM_021260406.1 | XP_021116065.1 | 19 - 20 | 4089 | 109 | ||
XM_021260407.1 | XP_021116066.1 | 17 - 18 | 3874 | 109 | ||
XM_021260408.1 | XP_021116067.1 | 18 - 19 | 4017 | 109 | ||
XM_021260409.1 | XP_021116068.1 | 18 - 19 | 3946 | 109 | ||
XM_021260410.1 | XP_021116069.1 | 19 - 20 | 4206 | 109 | ||
XM_021260411.1 | XP_021116070.1 | 19 - 20 | 4083 | 109 | ||
XM_021260412.1 | XP_021116071.1 | 17 - 18 | 3860 | 109 | ||
XM_021260413.1 | XP_021116072.1 | 15 - 16 | 2856 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
