Assay Details
Gene Symbol: Nsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847401.3 | XP_004847458.1 | 19 - 20 | 4409 | 71 | |
XM_013073021.2 | XP_012928475.1 | 20 - 21 | 4487 | 71 | ||
XM_021260406.1 | XP_021116065.1 | 24 - 25 | 4931 | 71 | ||
XM_021260407.1 | XP_021116066.1 | 22 - 23 | 4716 | 71 | ||
XM_021260408.1 | XP_021116067.1 | 23 - 24 | 4859 | 71 | ||
XM_021260409.1 | XP_021116068.1 | 23 - 24 | 4788 | 71 | ||
XM_021260410.1 | XP_021116069.1 | 24 - 25 | 5048 | 71 | ||
XM_021260411.1 | XP_021116070.1 | 24 - 25 | 4925 | 71 | ||
XM_021260412.1 | XP_021116071.1 | 22 - 23 | 4702 | 71 | ||
XM_021260413.1 | XP_021116072.1 | 20 - 21 | 3698 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
