Assay Details
Gene Symbol: Nsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847401.3 | XP_004847458.1 | 6 - 7 | 2446 | 79 | |
XM_013073021.2 | XP_012928475.1 | 7 - 8 | 2524 | 79 | ||
XM_021260406.1 | XP_021116065.1 | 11 - 12 | 2968 | 79 | ||
XM_021260407.1 | XP_021116066.1 | 9 - 10 | 2753 | 79 | ||
XM_021260408.1 | XP_021116067.1 | 10 - 11 | 2896 | 79 | ||
XM_021260409.1 | XP_021116068.1 | 10 - 11 | 2825 | 79 | ||
XM_021260410.1 | XP_021116069.1 | 11 - 12 | 3085 | 79 | ||
XM_021260411.1 | XP_021116070.1 | 11 - 12 | 2962 | 79 | ||
XM_021260412.1 | XP_021116071.1 | 9 - 10 | 2739 | 79 | ||
XM_021260413.1 | XP_021116072.1 | 7 - 8 | 1735 | 79 | ||
XM_021260414.1 | XP_021116073.1 | 7 - 8 | 1815 | 79 | ||
XM_021260415.1 | XP_021116074.1 | 7 - 8 | 1815 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
