Assay Details
Gene Symbol: Mob3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838371.2 | XP_004838428.1 | 2 - 3 | 1086 | 89 | |
XM_013069768.2 | XP_012925222.1 | 2 - 3 | 1092 | 89 | ||
XM_021252947.1 | XP_021108606.1 | 2 - 3 | 1102 | 89 | ||
XM_021252948.1 | XP_021108607.1 | 3 - 4 | 1268 | 89 | ||
XM_021252949.1 | XP_021108608.1 | 2 - 3 | 1090 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
