Assay Details
Gene Symbol: Tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845440.3 | XP_004845497.1 | 2 - 3 | 217 | 72 | |
XM_004845441.3 | XP_004845498.1 | 2 - 3 | 286 | 72 | ||
XM_004845442.3 | XP_004845499.1 | 2 - 3 | 210 | 72 | ||
XM_021258833.1 | XP_021114492.1 | 2 - 3 | 217 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
