Assay Details
Gene Symbol: Fam46c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853819.2 | XP_004853876.1 | 1 - 2 | 156 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004853819.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
