Assay Details
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004836374.3 | XP_004836431.1 | 5 - 6 | 501 | 64 | |
XM_004836375.3 | XP_004836432.1 | 6 - 7 | 721 | 64 | ||
XM_021239535.1 | XP_021095194.1 | 5 - 6 | 391 | 64 | ||
XM_021239545.1 | XP_021095204.1 | 4 - 5 | 480 | 64 | ||
XM_021239548.1 | XP_021095207.1 | 6 - 7 | 612 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
