Assay Details
Gene Symbol: Tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834833.2 | XP_004834890.1 | 4 - 5 | 879 | 79 | |
XM_004834835.2 | XP_004834892.1 | 3 - 4 | 1164 | 79 | ||
XM_021255930.1 | XP_021111589.1 | 4 - 5 | 594 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
