Assay Details
Gene Symbol: Mss51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013072780.2 | XP_012928234.1 | 3 - 4 | 1137 | 69 | |
XM_021259618.1 | XP_021115277.1 | 4 - 5 | 966 | 69 | ||
XM_021259619.1 | XP_021115278.1 | 4 - 5 | 1257 | 69 | ||
XM_021259620.1 | XP_021115279.1 | 5 - 6 | 1355 | 69 | ||
XM_021259621.1 | XP_021115280.1 | 3 - 4 | 1145 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
