Assay Details
| Gene Symbol: Cdkn2aipnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836572.2 | XP_004836629.1 | 1 - 2 | 311 | 71 | |
| XM_004836574.2 | XP_004836631.1 | 1 - 2 | 308 | 71 | ||
More Information
Set Membership: |
|

Cdkn2aipnl
Genomic Map