Assay Details
| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850626.3 | XP_004850683.2 | 2 - 3 | 678 | 57 | |
| XM_004850627.2 | XP_004850684.1 | 2 - 3 | 673 | 57 | ||
| XM_004850628.3 | XP_004850685.2 | 2 - 3 | 269 | 57 | ||
| XM_021263097.1 | XP_021118756.1 | 2 - 3 | 679 | 57 | ||
| XM_021263098.1 | XP_021118757.1 | 2 - 3 | 679 | 57 | ||
| XM_021263099.1 | XP_021118758.1 | 2 - 3 | 679 | 57 | ||
| XM_021263100.1 | XP_021118759.1 | 2 - 3 | 678 | 57 | ||
| XM_021263101.1 | XP_021118760.1 | 2 - 3 | 288 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021263101.1 transcripts.
Set Membership: |
|

Slc30a2
Genomic Map