Assay Details
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004844550.3 | XP_004844607.1 | 4 - 5 | 671 | 62 | |
XM_004844552.3 | XP_004844609.1 | 4 - 5 | 671 | 62 | ||
XM_021258186.1 | XP_021113845.1 | 3 - 4 | 666 | 62 | ||
XM_021258187.1 | XP_021113846.1 | 3 - 4 | 662 | 62 | ||
XM_021258188.1 | XP_021113847.1 | 3 - 4 | 672 | 62 | ||
XM_021258189.1 | XP_021113848.1 | 3 - 4 | 668 | 62 | ||
XM_021258190.1 | XP_021113849.1 | 3 - 4 | 672 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
