Assay Details
Gene Symbol: LOC101710289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862676.2 | XP_004862733.1 | 2 - 3 | 402 | 80 | |
XM_021242757.1 | XP_021098416.1 | 3 - 4 | 657 | 80 | ||
XM_021242758.1 | XP_021098417.1 | 3 - 4 | 665 | 80 | ||
XM_021242759.1 | XP_021098418.1 | 2 - 3 | 533 | 80 | ||
XM_021242760.1 | XP_021098419.1 | 3 - 4 | 671 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
