Assay Details
| Gene Symbol: Abcf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860374.3 | XP_004860431.1 | 13 - 14 | 1670 | 58 | |
| XM_021240954.1 | XP_021096613.1 | 13 - 14 | 1713 | 58 | ||
More Information
Set Membership: |
|

Abcf2
Genomic Map