Assay Details
Gene Symbol: Gpank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846981.3 | XP_004847038.1 | 1 - 2 | 1160 | 81 | |
XM_004846983.3 | XP_004847040.1 | 3 - 4 | 1161 | 81 | ||
XM_004846984.2 | XP_004847041.1 | 3 - 4 | 1074 | 81 | ||
XM_004846985.2 | XP_004847042.1 | 3 - 4 | 1125 | 81 | ||
XM_021260110.1 | XP_021115769.1 | 4 - 5 | 1134 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
