Assay Details
Gene Symbol: Serpinb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858449.2 | XP_004858506.1 | 6 - 7 | 868 | 80 | |
XM_021239269.1 | XP_021094928.1 | 9 - 10 | 1126 | 80 | ||
XM_021239270.1 | XP_021094929.1 | 8 - 9 | 1059 | 80 | ||
XM_021239271.1 | XP_021094930.1 | 8 - 9 | 1048 | 80 | ||
XM_021239272.1 | XP_021094931.1 | 8 - 9 | 1030 | 80 | ||
XM_021239273.1 | XP_021094932.1 | 7 - 8 | 939 | 80 | ||
XM_021239274.1 | XP_021094933.1 | 7 - 8 | 948 | 80 | ||
XM_021239276.1 | XP_021094935.1 | 7 - 8 | 981 | 80 | ||
XM_021239277.1 | XP_021094936.1 | 6 - 7 | 870 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
