Assay Details
Gene Symbol: Pex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842031.3 | XP_004842088.1 | 2 - 3 | 290 | 89 | |
XM_004842032.2 | XP_004842089.1 | 3 - 4 | 405 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004842031.3, XM_004842032.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
