Assay Details
Gene Symbol: Prkra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004868141.3 | XP_004868198.1 | 2 - 3 | 437 | 65 | |
XM_021247137.1 | XP_021102796.1 | 1 - 2 | 168 | 65 | ||
XM_021247138.1 | XP_021102797.1 | 2 - 3 | 361 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021247137.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
