Assay Details
Gene Symbol: Wdr86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860367.3 | XP_004860424.1 | 3 - 4 | 1316 | 80 | |
XM_004860368.3 | XP_004860425.1 | 3 - 4 | 1238 | 80 | ||
XM_004860369.3 | XP_004860426.1 | 3 - 4 | 1317 | 80 | ||
XM_021240911.1 | XP_021096570.1 | 3 - 4 | 1319 | 80 | ||
XM_021240912.1 | XP_021096571.1 | 3 - 4 | 1242 | 80 | ||
XM_021240913.1 | XP_021096572.1 | 3 - 4 | 6618 | 80 | ||
XM_021240914.1 | XP_021096573.1 | 3 - 4 | 1317 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
