Assay Details
| Gene Symbol: Scgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847585.2 | XP_004847642.1 | 10 - 11 | 906 | 77 | |
| XM_021260540.1 | XP_021116199.1 | 9 - 10 | 828 | 77 | ||
| XM_021260541.1 | XP_021116200.1 | 9 - 10 | 816 | 77 | ||
More Information
Set Membership: |
|

Scgn
Genomic Map