Assay Details
| Gene Symbol: Pacsin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845446.3 | XP_004845503.1 | 4 - 5 | 691 | 58 | |
| XM_004845447.3 | XP_004845504.1 | 4 - 5 | 641 | 58 | ||
| XM_004845448.3 | XP_004845505.1 | 4 - 5 | 554 | 58 | ||
| XM_004845449.3 | XP_004845506.1 | 4 - 5 | 608 | 58 | ||
| XM_013072524.2 | XP_012927978.1 | 4 - 5 | 641 | 58 | ||
| XM_013072525.2 | XP_012927979.1 | 4 - 5 | 641 | 58 | ||
| XM_013072526.2 | XP_012927980.1 | 4 - 5 | 641 | 58 | ||
| XM_021258848.1 | XP_021114507.1 | 4 - 5 | 751 | 58 | ||
| XM_021258849.1 | XP_021114508.1 | 3 - 4 | 506 | 58 | ||
More Information
Set Membership: |
|

Pacsin2
Genomic Map