Assay Details
Gene Symbol: Lzic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004863633.2 | XP_004863690.1 | 5 - 6 | 759 | 61 | |
XM_004863636.2 | XP_004863693.1 | 4 - 5 | 520 | 61 | ||
XM_021243407.1 | XP_021099066.1 | 6 - 7 | 811 | 61 | ||
XM_021243408.1 | XP_021099067.1 | 5 - 6 | 558 | 61 | ||
XM_021243409.1 | XP_021099068.1 | 4 - 5 | 722 | 61 | ||
XM_021243410.1 | XP_021099069.1 | 5 - 6 | 558 | 61 | ||
XM_021243411.1 | XP_021099070.1 | 5 - 6 | 498 | 61 | ||
XM_021243412.1 | XP_021099071.1 | 5 - 6 | 463 | 61 | ||
XM_021243413.1 | XP_021099072.1 | 4 - 5 | 721 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
