Assay Details
| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852918.2 | XP_004852975.1 | 3 - 4 | 591 | 81 | |
| XM_021264921.1 | XP_021120580.1 | 4 - 5 | 690 | 81 | ||
| XM_021264922.1 | XP_021120581.1 | 3 - 4 | 579 | 81 | ||
| XM_021264923.1 | XP_021120582.1 | 4 - 5 | 690 | 81 | ||
| XM_021264924.1 | XP_021120583.1 | 4 - 5 | 690 | 81 | ||
| XM_021264925.1 | XP_021120584.1 | 3 - 4 | 288 | 81 | ||
More Information
Set Membership: |
|

Cbr4
Genomic Map