Assay Details
Gene Symbol: Slc39a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848300.2 | XP_004848357.1 | 2 - 3 | 708 | 59 | |
XM_004848301.2 | XP_004848358.1 | 2 - 3 | 708 | 59 | ||
XM_021261241.1 | XP_021116900.1 | 2 - 3 | 708 | 59 | ||
XM_021261242.1 | XP_021116901.1 | 2 - 3 | 518 | 59 | ||
XM_021261243.1 | XP_021116902.1 | 2 - 3 | 381 | 59 | ||
XM_021261244.1 | XP_021116903.1 | 2 - 3 | 353 | 59 | ||
XM_021261245.1 | XP_021116904.1 | 2 - 3 | 476 | 59 | ||
XM_021261246.1 | XP_021116905.1 | 2 - 3 | 296 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
