Assay Details
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866877.3 | XP_004866934.1 | 4 - 5 | 553 | 61 | |
XM_004866879.3 | XP_004866936.1 | 3 - 4 | 343 | 61 | ||
XM_004866880.3 | XP_004866937.1 | 4 - 5 | 592 | 61 | ||
XM_021245841.1 | XP_021101500.1 | 4 - 5 | 561 | 61 | ||
XM_021245842.1 | XP_021101501.1 | 4 - 5 | 630 | 61 | ||
XM_021245843.1 | XP_021101502.1 | 4 - 5 | 602 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
