Assay Details
| Gene Symbol: Keap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865501.3 | XP_004865558.1 | 3 - 4 | 1638 | 63 | |
| XM_004865502.3 | XP_004865559.1 | 3 - 4 | 1529 | 63 | ||
| XM_004865503.3 | XP_004865560.1 | 3 - 4 | 1623 | 63 | ||
More Information
Set Membership: |
|

Keap1
Genomic Map