Assay Details
Gene Symbol: Tarbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068476.2 | XP_012923930.1 | 8 - 9 | 1067 | 54 | |
XM_021250658.1 | XP_021106317.1 | 8 - 9 | 1094 | 54 | ||
XM_021250659.1 | XP_021106318.1 | 8 - 9 | 1058 | 54 | ||
XM_021250660.1 | XP_021106319.1 | 7 - 8 | 974 | 54 | ||
XM_021250661.1 | XP_021106320.1 | 7 - 8 | 947 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
