Assay Details
Gene Symbol: Triqk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850092.2 | XP_004850149.1 | 3 - 4 | 306 | 72 | |
XM_004850093.2 | XP_004850150.1 | 4 - 5 | 467 | 72 | ||
XM_021262626.1 | XP_021118285.1 | 4 - 5 | 741 | 72 | ||
XM_021262627.1 | XP_021118286.1 | 4 - 5 | 464 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
