Assay Details
Gene Symbol: Pag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021255990.1 | XP_021111649.1 | 6 - 7 | 1126 | 83 | |
XM_021255991.1 | XP_021111650.1 | 7 - 8 | 1259 | 83 | ||
XM_021255992.1 | XP_021111651.1 | 6 - 7 | 1112 | 83 | ||
XM_021255993.1 | XP_021111652.1 | 5 - 6 | 502 | 83 | ||
XM_021255994.1 | XP_021111653.1 | 6 - 7 | 1132 | 83 | ||
XM_021255995.1 | XP_021111654.1 | 6 - 7 | 1227 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
