Assay Details
Gene Symbol: Poc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834194.3 | XP_004834251.3 | 7 - 8 | 1173 | 63 | |
XM_021241199.1 | XP_021096858.1 | 7 - 8 | 1176 | 63 | ||
XM_021241238.1 | XP_021096897.1 | 7 - 8 | 1120 | 63 | ||
XM_021241281.1 | XP_021096940.1 | 7 - 8 | 1062 | 63 | ||
XM_021241366.1 | XP_021097025.1 | 7 - 8 | 816 | 63 | ||
XM_021241403.1 | XP_021097062.1 | 7 - 8 | 1171 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
