Assay Details
Gene Symbol: LOC101707725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021249563.1 | XP_021105222.1 | 6 - 7 | 1305 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
