Assay Details
| Gene Symbol: Ccnq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874928.2 | XP_004874985.1 | 2 - 3 | 394 | 95 | |
| XM_004874933.3 | XP_004874990.1 | 2 - 3 | 394 | 95 | ||
| XM_013069375.2 | XP_012924829.1 | 2 - 3 | 394 | 95 | ||
| XM_013069376.2 | XP_012924830.1 | 2 - 3 | 394 | 95 | ||
| XM_021252329.1 | XP_021107988.1 | 2 - 3 | 394 | 95 | ||
More Information
Set Membership: |
|

Ccnq
Genomic Map