Assay Details
| Gene Symbol: LOC101713959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872496.3 | XP_004872553.1 | 2 - 3 | 3662 | 100 | |
| XM_021250407.1 | XP_021106066.1 | 10 - 11 | 2273 | 100 | ||
| XM_021250408.1 | XP_021106067.1 | 11 - 12 | 2309 | 100 | ||
| XM_021250409.1 | XP_021106068.1 | 11 - 12 | 2327 | 100 | ||
| XM_021250410.1 | XP_021106069.1 | 11 - 12 | 2321 | 100 | ||
| XM_021250411.1 | XP_021106070.1 | 10 - 11 | 2243 | 100 | ||
More Information
Set Membership: |
|

LOC101713959
Genomic Map