Assay Details
Gene Symbol: Gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850677.3 | XP_004850734.1 | 4 - 5 | 1088 | 68 | |
XM_004850678.3 | XP_004850735.1 | 4 - 5 | 1019 | 68 | ||
XM_021263129.1 | XP_021118788.1 | 4 - 5 | 1100 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
