Assay Details
Gene Symbol: Nmrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865871.3 | XP_004865928.1 | 4 - 5 | 455 | 76 | |
XM_013065944.2 | XP_012921398.1 | 4 - 5 | 470 | 76 | ||
XM_021244942.1 | XP_021100601.1 | 4 - 5 | 564 | 76 | ||
XM_021244943.1 | XP_021100602.1 | 4 - 5 | 531 | 76 | ||
XM_021244944.1 | XP_021100603.1 | 4 - 5 | 525 | 76 | ||
XM_021244945.1 | XP_021100604.1 | 4 - 5 | 470 | 76 | ||
XM_021244946.1 | XP_021100605.1 | 3 - 4 | 457 | 76 | ||
XM_021244947.1 | XP_021100606.1 | 4 - 5 | 455 | 76 | ||
XM_021244948.1 | XP_021100607.1 | 3 - 4 | 607 | 76 | ||
XM_021244949.1 | XP_021100608.1 | 3 - 4 | 324 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
