Assay Details
Gene Symbol: LOC101726271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870750.3 | XP_004870807.1 | 1 - 2 | 184 | 91 | |
XM_021248989.1 | XP_021104648.1 | 1 - 2 | 184 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
