Assay Details
Gene Symbol: Rcc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840009.2 | XP_004840066.1 | 2 - 3 | 533 | 70 | |
XM_013070472.2 | XP_012925926.1 | 2 - 3 | 539 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
