Assay Details
| Gene Symbol: Brms1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867551.3 | XP_004867608.1 | 8 - 9 | 916 | 79 | |
| XM_004867552.3 | XP_004867609.1 | 8 - 9 | 918 | 79 | ||
| XM_021246635.1 | XP_021102294.1 | 8 - 9 | 919 | 79 | ||
| XM_021246636.1 | XP_021102295.1 | 8 - 9 | 919 | 79 | ||
| XM_021246637.1 | XP_021102296.1 | 9 - 10 | 778 | 79 | ||
More Information
Set Membership: |
|

Brms1l
Genomic Map