Assay Details
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013071006.2 | XP_012926460.1 | 10 - 11 | 1115 | 58 | |
XM_021255611.1 | XP_021111270.1 | 9 - 10 | 1261 | 58 | ||
XM_021255612.1 | XP_021111271.1 | 10 - 11 | 1117 | 58 | ||
XM_021255613.1 | XP_021111272.1 | 9 - 10 | 1261 | 58 | ||
XM_021255615.1 | XP_021111274.1 | 6 - 7 | 922 | 58 | ||
XM_021255616.1 | XP_021111275.1 | 6 - 7 | 874 | 58 | ||
XM_021255617.1 | XP_021111276.1 | 5 - 6 | 838 | 58 | ||
XM_021255618.1 | XP_021111277.1 | 9 - 10 | 1012 | 58 | ||
XM_021255619.1 | XP_021111278.1 | 10 - 11 | 1130 | 58 | ||
XM_021255620.1 | XP_021111279.1 | 9 - 10 | 1007 | 58 | ||
XM_021255621.1 | XP_021111280.1 | 10 - 11 | 1125 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
